missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Phenylethyl)piperazine, 98%

Product Code. 11496237
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496237 10 g 10g
11406247 50 g 50g
11416247 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496237 Supplier Thermo Scientific Alfa Aesar Supplier No. B25158.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5321-49-3 | C12H18N2 | 190.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5321-49-3
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00040740
InChI Key LKUAPSRIYZLAAO-UHFFFAOYSA-N
Synonym 1-2-phenylethyl piperazine, 1-phenethylpiperazine, 1-phenethyl-piperazine, 1-2-phenylethyl-piperazine, 1-phenylethylpiperazine, 1-phenethyl piperazine, n-2-phenylethyl piperazine, piperazine, 1-2-phenylethyl, 2-phenylethyl piperazine, l-phenethylpiperazine
PubChem CID 79214
IUPAC Name 1-(2-phenylethyl)piperazine
SMILES C1CN(CCN1)CCC2=CC=CC=C2

Specifications

Density 1.01
Boiling Point 276°C
Flash Point >110°C (230°F)
Refractive Index 1.543
Quantity 10 g
UN Number UN2735
Beilstein 145893
Sensitivity Air Sensitive
Formula Weight 190.29
Percent Purity 98%
Chemical Name or Material 1-(2-Phenylethyl)piperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.