missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, TRC

Product Code. 30425523
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425523 50 g 50g
30419638 10 g 10g
30420428 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425523 Supplier Toronto Research Chemicals Supplier No. TRCP76031050G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Propanediol
CAS 57-55-6
Purity Grade Notes GC
Molecular Formula C3 H8 O2
Quantity 50 g
Synonym Propylene glycol, (RS)-propane-1,2-diol
InChI Formula InChI=1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3
SMILES CC(O)CO
IUPAC Name propane-1,2-diol
Molecular Weight (g/mol) 76.09
Formula Weight 76.0524
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.