missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyrimidinyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™

Product Code. 11489530
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489530 1 g 1g
11479530 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489530 Supplier Thermo Scientific Alfa Aesar Supplier No. H50351.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303144-44-7
Molecular Formula C10H12N3O2
Molecular Weight (g/mol) 206.23
MDL Number MFCD00664474
InChI Key DYVXURZASBPYFR-UHFFFAOYSA-M
Synonym 1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl
PubChem CID 2767267
IUPAC Name 1-pyrimidin-2-ylpiperidine-4-carboxylic acid
SMILES [O-]C(=O)C1CCN(CC1)C1=NC=CC=N1

Specifications

Quantity 1 g
Formula Weight 207.23
Percent Purity 95%
Chemical Name or Material 1-(2-Pyrimidinyl)piperidine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.