missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydrofuroyl)piperazine, 97%

Product Code. 11389475
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11389475 25 g 25g
11379475 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11389475

Brand: Thermo Scientific Alfa Aesar L18704.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63074-07-7 | C9H16N2O2 | 184.239 g/mol

An intermediate of terazosin hydrochloride. It has also been used as a reactant for the preparation of pyrazol-3-propanoic acid derivatives as inhibitors of leukotriene biosynthesis in human neutrophils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An intermediate of terazosin hydrochloride. It has also been used as a reactant for the preparation of pyrazol-3-propanoic acid derivatives as inhibitors of leukotriene biosynthesis in human neutrophils.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63074-07-7
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.239
MDL Number MFCD00190247
InChI Key UKESBLFBQANJHH-UHFFFAOYSA-N
Synonym 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone
PubChem CID 2734641
IUPAC Name oxolan-2-yl(piperazin-1-yl)methanone
SMILES C1CC(OC1)C(=O)N2CCNCC2

Specifications

Boiling Point 120°C to 125°C (0.2 mmHg)
Quantity 25 g
Beilstein 880578
Solubility Information Fully miscible in water.
Formula Weight 184.24
Percent Purity 97%
Chemical Name or Material 1-(2-Tetrahydrofuroyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.