missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester, 95%

Product Code. 15498065
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498065 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498065 Supplier Thermo Scientific Alfa Aesar Supplier No. H32369.ME
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1029684-37-4 | C14H23BN2O3 | 278.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1029684-37-4
Melting Point 129°C to 133°C
Molecular Formula C14H23BN2O3
MDL Number MFCD11100960
Quantity 500 mg
Synonym 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 1-tetrahydro-2h-pyran-2-yl-1h-imidazole-5-boronic acid pinacol ester, 1-2-tetrahydropyranyl-1h-imidazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 1-tetrahydro-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-imidazole, 1-tetrahydropyran-2-yl-1h-imidazole-5-boronic ac, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole, 1-tetrahydro-pyran-2-yl-1h-imidazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-imidazole-5-boronic acid pinacol ester
Solubility Information Slightly soluble in water.
InChI Key BVIWGBKKGPHNAB-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=CN2C3CCCCO3
IUPAC Name 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazole
Molecular Weight (g/mol) 278.159
PubChem CID 24880008
Formula Weight 278.16
Percent Purity 95%
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.