missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 11441710
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441710 1 g 1g
15417655 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441710 Supplier Thermo Scientific Alfa Aesar Supplier No. H27036.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 903550-26-5 | Molecular Formula: C14H23BN2O3 | Molecular Weight: 278.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 903550-26-5
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.159
MDL Number MFCD09037501
InChI Key ZZRFDLHBMBHJTI-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-2-tetrahydropyranyl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-y-1h-pyrazole, 1-tetrahydropyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole
PubChem CID 11587208
IUPAC Name 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2C3CCCCO3

Specifications

Quantity 1 g
Formula Weight 278.16
Percent Purity 95%
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.