missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thiazolylazo)-2-naphthol, 98%

Product Code. 11486916
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486916 1 g 1g
11496916 5 g 5g
11406926 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486916 Supplier Thermo Scientific Alfa Aesar Supplier No. B24011.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1147-56-4 | C13H9N3OS | 255.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1147-56-4
Molecular Formula C13H9N3OS
Molecular Weight (g/mol) 255.30
MDL Number MFCD00021611
InChI Key GCBAKGGJQPMJLL-QINSGFPZSA-N
Synonym 1-2-thiazolylazo-2-naphthol, 1-2-thiazoylazo-2-naphthol, 2-naphthol, 1-2-thiazolylazo, 2-naphthalenol, 1-2-thiazolylazo, 1z-1-1,3-thiazol-2-ylhydrazinylidene naphthalen-2-one, 1-1,3-thiazol-2-yldiazenyl-2-naphthol, 1z-1-2-thiazolylhydrazinylidene-2-naphthalenone, 1-thiazol-2-ylazo-2-naphthol, 2-naphthalenol, 1-2-2-thiazolyl diazenyl, iomxcgdxeudzak-foclmdbbsa-n
PubChem CID 6308684
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=NC=CS1

Specifications

Melting Point 138°C to 140°C
Quantity 1 g
Beilstein 223783
IUPAC Name (1Z)-1-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Formula Weight 255.3
Percent Purity 98%
Chemical Name or Material 1-(2-Thiazolylazo)-2-naphthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.