missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thienyl)ethylamine, tech. 90%

Product Code. 15473357
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473357 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15473357 Supplier Thermo Scientific Alfa Aesar Supplier No. H26052.ME
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 6309-16-6 | C6H10NS | 128.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6309-16-6
Molecular Formula C6H10NS
Refractive Index 1.5475
MDL Number MFCD02734311
Quantity 500 mg
UN Number UN2735
Sensitivity Air Sensitive
Synonym 1-thiophen-2-yl ethanamine, 1-2-thienyl ethylamine, 1-thiophen-2-yl-ethylamine, 1-thiophen-2-yl ethan-1-amine, 1-thien-2-ylethanamine, 1-2-thienyl ethanamine, 1-2-thienyl ethanamin, 1-thiophen-2-ylethylamine, 1-thien-2-yl ethylamine, 2-1-aminoethyl thiophene
InChI Key LYJBVRVJQXVVPI-RXMQYKEDSA-O
SMILES C[C@@H]([NH3+])C1=CC=CS1
IUPAC Name 1-thiophen-2-ylethanamine
Molecular Weight (g/mol) 128.21
PubChem CID 238233
Formula Weight 127.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-(2-Thienyl)ethylamine
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Thienyl)ethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1-(2-Thienyl)ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.