missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Trifluoromethyl)phenyl]pyrrole, 98%, Thermo Scientific™

Product Code. 15477067
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15477067 25 g 25g
15457067 1 g 1g
15467067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15477067 Supplier Thermo Scientific Alfa Aesar Supplier No. H34139.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89108-30-5
Molecular Formula C11H8F3N
Molecular Weight (g/mol) 211.187
MDL Number MFCD02665223
InChI Key IUMZRHODYBOXLA-UHFFFAOYSA-N
PubChem CID 11367822
IUPAC Name 1-[2-(trifluoromethyl)phenyl]pyrrole
SMILES C1=CC=C(C(=C1)C(F)(F)F)N2C=CC=C2

Specifications

Boiling Point 62°C (1.13 mmHg)
Quantity 25 g
Synonym 1h-pyrrole, 1-2-trifluoromethyl phenyl, 1-2-trifluoromethyl phenyl pyrrole, 1-2-trifluoromethyl phenyl-1h-pyrrole, acmc-20lhrq, 1-2-trifluoromethyl phenyl-1 h-pyrrole
Formula Weight 211.19
Percent Purity 98%
Chemical Name or Material 1-[2-(Trifluoromethyl)phenyl]pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.