missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Trimethylsilylethoxy)methyl-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 11496070
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496070 1 g 1g
15495115 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496070 Supplier Thermo Scientific Alfa Aesar Supplier No. H27043.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 903550-12-9
Molecular Formula C15H29BN2O3Si
Molecular Weight (g/mol) 324.303
MDL Number MFCD09037502
InChI Key FVCZMGOMGHMAIX-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl-1h-pyrazole, 1-2-trimethylsilylethoxy methylpyrazole-5-boronic acid, pinacol ester, 1-2-trimethylsilanylethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxy methyl-1h-pyrazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl pyrazole, 1-2-trimethylsilyl ethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxyl methyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilyl ethoxymethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-pyrazole, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-2-trimethylsilanyl-ethoxymethyl-1h-pyrazole
PubChem CID 44755201
IUPAC Name trimethyl-[2-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methoxy]ethyl]silane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2COCC[Si](C)(C)C

Specifications

Quantity 1 g
Formula Weight 324.31
Percent Purity 95%
Chemical Name or Material 1-(2-Trimethylsilylethoxy)methyl-1H-pyrazole-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.