missing translation for 'onlineSavingsMsg'
Learn More

1-(2H-1,3-benzodioxol-5-yl)propan-2-ol, TRC

Product Code. 30132762
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132762 100 mg 100mg
30010799 10 mg 10mg
30010398 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132762 Supplier Toronto Research Chemicals Supplier No. TRCB115030100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6974-61-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
Synonym 1,3-Benzodioxole-5-ethanol, α-methyl-, Phenethyl alcohol, α-methyl-3,4-(methylenedioxy)- (6CI,8CI), α-Methyl-1,3-benzodioxole-5-ethanol, 1-(Benzo[1,3]dioxol-5-yl)-2-propanol, 1-[3,4-(Methylenedioxy)phenyl]-2-propanol, 3-(1,3-Benzodioxol-5-yl)propan-2-ol, NSC 15666, 1-(3,4-Methylenedioxyphenyl)propan-2-ol
IUPAC Name 1-(1,3-benzodioxol-5-yl)propan-2-ol
SMILES CC(O)Cc1ccc2OCOc2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12O3/c1-7(11)4-8-2-3-9-10(5-8)13-6-12-9/h2-3,5,7,11H,4,6H2,1H3
Formula Weight 180.0786
Chemical Name or Material 1-(2H-1,3-benzodioxol-5-yl)propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.