missing translation for 'onlineSavingsMsg'
Learn More

1,3,2-Dioxathiolane 2,2-dioxide, 96%, Thermo Scientific Chemicals

Product Code. 10706895 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10706895 1 g Glass bottle 1g
10401304 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10706895 Supplier Thermo Scientific Acros Supplier No. 426630010
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-53-3
Molecular Formula C2H4O4S
Molecular Weight (g/mol) 124.12
MDL Number MFCD00221769
InChI Key ZPFAVCIQZKRBGF-UHFFFAOYSA-N
Synonym ethylenesulfate, ethylene sulfate, ethylene glycol, cyclic sulfate, glycol sulfate, 1,2-ethylene sulfate, 1,3,2-dioxathiolane, 2,2-dioxide, sulfuric acid, cyclic ethylene ester, 1,3,2-dioxathiolane-2,2-dioxide, 1,3,2??-dioxathiolane-2,2-dione, ethosulfate
PubChem CID 14075
IUPAC Name 1,3,2-dioxathiolane 2,2-dioxide
SMILES C1COS(=O)(=O)O1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 95°C to 97°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 124.12
Percent Purity 96%
Chemical Name or Material 1,3,2-Dioxathiolane 2,2-dioxide
compliance-icons
Product Identifier
  • 1,3,2-Dioxathiolane 2,2-dioxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.