missing translation for 'onlineSavingsMsg'
Learn More

1,3,2-Dioxathiolane 2,2-Dioxide, TRC

Product Code. 30377351
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377351 1 g
30369907 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30377351 Supplier Toronto Research Chemicals Supplier No. TRCD4868651G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-53-3
Molecular Formula C2 H4 O4 S
Molecular Weight (g/mol) 124.12
Synonym Ethylene Glycol Cyclic Sulfate, 1,2-Ethylene Sulfate, Ethylene Sulfate, NSC 526594
IUPAC Name 1,3,2-dioxathiolane 2,2-dioxide
SMILES O=S1(=O)OCCO1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H4O4S/c3-7(4)5-1-2-6-7/h1-2H2
Formula Weight 123.983
Chemical Name or Material 1,3,2-Dioxathiolane 2,2-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.