missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dichlorobenzyl)piperazine, 97%

Product Code. 15442008
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442008 25 g 25g
15422008 1 g 1g
15432008 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442008 Supplier Thermo Scientific Alfa Aesar Supplier No. H55685.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 55513-17-2 | C11H14Cl2N2 | 245.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55513-17-2
Molecular Formula C11H14Cl2N2
Molecular Weight (g/mol) 245.147
MDL Number MFCD03407488
InChI Key PNDSYXGJCWKNFG-UHFFFAOYSA-N
Synonym 1-3,4-dichlorobenzyl piperazine, 1-3,4-dichlorophenyl methyl piperazine, piperazine, 1-3,4-dichlorophenyl methyl, 1-3,4-dichloro-benzyl-piperazine, acmc-20apc5, 3,4-dichlorobenzylpiperazine, 1-3,4-dichlorobenyl piperazine, 3,4-dichlorophenyl methyl piperazine
PubChem CID 2758074
IUPAC Name 1-[(3,4-dichlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC(=C(C=C2)Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.225 g/mL
Boiling Point 124°C to 128°C (0.02 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 245.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3,4-Dichlorobenzyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.