missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dichlorophenyl)-1-propanone, 99%, ACROS Organics™

Product Code. 10215520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10215520 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10215520 Supplier Thermo Scientific Acros Supplier No. 179570050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6582-42-9
CAS Min % 98.5
CAS Max % 100.0
Melting Point 41°C to 45°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H8Cl2O
Linear Formula Cl2C6H3COC2H5
MDL Number MFCD00000554
Quantity 5 g
Synonym 3',4'-dichloropropiophenone, 1-3,4-dichlorophenyl propan-1-one, 1-3,4-dichlorophenyl-1-propanone, 1-propanone, 1-3,4-dichlorophenyl, 1-propanone,1-3,4-dichlorophenyl, enamine_003892, acmc-1b5az, 3,4-dichloro propiophenone, 1-3,4-dichlorophenyl-1-propanone #
InChI Key AJUAASWQUWIMHM-UHFFFAOYSA-N
SMILES CC(=O)CC1=CC=C(Cl)C(Cl)=C1
IUPAC Name 1-(3,4-dichlorophenyl)propan-2-one
Molecular Weight (g/mol) 203.06
PubChem CID 599598
Formula Weight 203.07
Percent Purity ≥98.5%
Physical Form Low Melting Solid
Chemical Name or Material 1-(3, 4-Dichlorophenyl)-1-propanone, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.