missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dihydro-7-hydroxy-6-methoxy-2(1H)-isoquinolinyl)-1-propanone, TRC

Product Code. 30369979
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369979 250 mg 250mg
30384161 50 mg -
30379690 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369979 Supplier Toronto Research Chemicals Supplier No. TRCD444995250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1032822-42-6
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.28
SMILES CCC(=O)N1CCc2cc(OC)c(O)cc2C1

Tekniske data

Quantity 250 mg
InChI Formula InChI=1S/C13H17NO3/c1-3-13(16)14-5-4-9-7-12(17-2)11(15)6-10(9)8-14/h6-7,15H,3-5,8H2,1-2H3
Formula Weight 235.1208
Chemical Name or Material 1-(3,4-Dihydro-7-hydroxy-6-methoxy-2(1H)-isoquinolinyl)-1-propanone
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.