missing translation for 'onlineSavingsMsg'
Learn More

1,3,5,7-Cyclooctatetraene, 98%, stab. with 0.1% Hydroquinone

Product Code. 11349779
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349779 5 g 5g
11359779 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349779 Supplier Thermo Scientific Alfa Aesar Supplier No. B22636.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-20-9 | C8H8 | 104.15 g/mol

1,3,5,7-Cyclooctatetraene is used in the synthesis of highly organic film for silicon surfaces to improve its chemical and physical properties. It is also used in liquid state organic dye lasers, as triplet state quencher to reduce dye blinking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5,7-Cyclooctatetraene is used in the synthesis of highly organic film for silicon surfaces to improve its chemical and physical properties. It is also used in liquid state organic dye lasers, as triplet state quencher to reduce dye blinking.

Solubility
Not miscible or difficult to mix in water.

Notes
Air & light sensitive. Store at --20°C. Store away from oxidizing agent, air, heat, ultra violet radiation, free radical intiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-20-9
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00004161
InChI Key KDUIUFJBNGTBMD-DLMDZQPMSA-N
Synonym 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
PubChem CID 637866
IUPAC Name cyclooctatetraene
SMILES C1=C/C=C\C=C/C=C\1

Specifications

Melting Point -5°C
Density 0.925
Boiling Point 142°C to 143°C
Flash Point 20°C (68°F)
Odor Unpleasant
Refractive Index 1.537
Quantity 5 g
UN Number UN2358
Beilstein 2496800
Sensitivity Air and light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 104.15
Percent Purity 98%
Chemical Name or Material 1,3,5,7-Cyclooctatetraene, stabilized with 0.1% Hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.