missing translation for 'onlineSavingsMsg'
Learn More

1,3,5,7-Cyclooctatetraene, 98%, stabilized

Product Code. 10194120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10194120 2.5 g Glass bottle 2.5g
10637934 250 mg Glass bottle 250mg
10413645 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10194120 Supplier Thermo Scientific Acros Supplier No. 160570025
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-20-9 | C8H8 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-20-9
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00004161
InChI Key KDUIUFJBNGTBMD-DLMDZQPMSA-N
Synonym 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
PubChem CID 637866
IUPAC Name cyclooctatetraene
SMILES C1=C/C=C\C=C/C=C\1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -27°C
Color Brown-Yellow to Yellow
Density 0.9200g/mL
Boiling Point 142°C to 143°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5365 to 1.5385
Quantity 2.5 g
Beilstein 05, I, 228
Merck Index 15, 2731
Specific Gravity 0.92
Solubility Information Solubility in water: immiscible
Stabilizer 0.1% hydroquinone
Formula Weight 104.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3,5,7-Cyclooctatetraene, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3,5,7-Cyclooctatetraene, stabilized
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1,3,5,7-Cyclooctatetraene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.