missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid, 98%

Product Code. 11458496
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
Unit Size:
100g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458496 50 g 50g
11324599 100 g 100g
11468496 250 g 250g
11334599 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11458496 Supplier Thermo Scientific Alfa Aesar Supplier No. A15947.18
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 554-95-0 | C9H6O6 | 210.14 g/mol

1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

Solubility
Soluble in water, ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-95-0
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002517
InChI Key QMKYBPDZANOJGF-UHFFFAOYSA-N
Synonym trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
PubChem CID 11138
ChEBI CHEBI:46032
IUPAC Name benzene-1,3,5-tricarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

Melting Point 374°C to 376°C
Flash Point 328°C (622°F)
Quantity 50 g
Beilstein 2053080
Solubility Information Soluble in water,ethanol and methanol.
Formula Weight 210.14
Percent Purity 98%
Chemical Name or Material 1,3,5-Benzenetricarboxylic acid
compliance-icons
Product Identifier
  • 1,3,5-Benzenetricarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,3,5-Benzenetricarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.