Learn More
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 380.0°C |
| Flash Point | >250°C |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Plastic bottle |
| Linear Formula | C6H3(CO2H)3 |
| Quantity | 50 g |
| Beilstein | 09, 978 |
| Solubility Information | Solubility in water: 2.6g/L (25°C)- 7.9g/L (50°C). Other solubilities: 1.24g/100g ethyl ether- 8g/100g methanol,0.46g/100g acetic acid- >20.00g/100g thf (25°C),1.88g/100g acetone- <0.01g/100g xylene (25°C),<0.01g/1 |
| Mostrar más |
Seguridad y manipulación
- 1,3,5-Benzenetricarboxylic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.