missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid, 98%

Product Code. 10730585 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
500g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10730585 500 g Plastic bottle 500g
10163581 5 g Glass Bottle 5g
10253661 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10730585 Supplier Thermo Scientific Acros Supplier No. 105355000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1, 3, 5-Benzenetricarboxylic acid, 98%, C9H6O6, CAS Number-554-95-0, 1,3,5-benzene tricarboxylic acid, 1,3,5-benzenetricarboxylic acid, 1,3,5-tricarboxybenzene, 5-carboxyisophthalic acid, chembl77562, ou36oo5mtn, trimesic acid, trimesinic acid, trimesitinic acid, unii-ou36oo5mtn, 500g | CAS: 554-95-0 | C9H6O6 | 210.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-95-0
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002517
InChI Key QMKYBPDZANOJGF-UHFFFAOYSA-N
Synonym trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
PubChem CID 11138
ChEBI CHEBI:46032
IUPAC Name benzene-1,3,5-tricarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 380.0°C
Flash Point >250°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H3(CO2H)3
Quantity 500 g
Beilstein 09, 978
Solubility Information Solubility in water: 2.6g/L (25°C)- 7.9g/L (50°C). Other solubilities: 1.24g/100g ethyl ether- 8g/100g methanol,0.46g/100g acetic acid- >20.00g/100g thf (25°C),1.88g/100g acetone- <0.01g/100g xylene (25°C),<0.01g/1
Formula Weight 210.14
Percent Purity 98%
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1,3,5-Benzenetricarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.