missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Benzenetricarboxylic acid chloride, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 34.5°C to 36°C |
| Color | Colorless to Yellow |
| Boiling Point | 180°C (16.0 mmHg) |
| Flash Point | >112°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5 to 102.5% (ex Cl) (Argentometry) |
| Packaging | Glass bottle |
| Linear Formula | C6H3(COCl)3 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 1,3,5-Benzenetricarboxylic acid chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur