missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Cycloheptatriene, TRC

Product Code. 30365655
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
Unit Size:
10mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365655 50 mL 50mL
30358032 25 mL 25mL
30352156 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365655 Supplier Toronto Research Chemicals Supplier No. TRCC98786250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-25-2
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
Synonym Cycloheptatriene, Tropiliden, Tropilidene
IUPAC Name cyclohepta-1,3,5-triene
SMILES C1C=CC=CC=C1

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C7H8/c1-2-4-6-7-5-3-1/h1-6H,7H2
Formula Weight 92.0626
Chemical Name or Material 1,3,5-Cycloheptatriene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.