missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Difluoro-2-hydroxyphenyl)ethan-1-one, Thermo Scientific™

Product Code. 10272772
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10272772 10 g Amber Glass Bottle 10g
10262772 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10272772 Supplier Thermo Scientific Maybridge Supplier No. BTB04131EA
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 140675-42-9
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042476
InChI Key MCDJUVXLLXTCFP-UHFFFAOYSA-N
Synonym 1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone
PubChem CID 2736976
IUPAC Name 1-(3,5-difluoro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 1-(3, 5-Difluoro-2-hydroxyphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.