missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri(1-naphthyl)benzene, TRC

Product Code. 30451682
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451682 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451682 Supplier Toronto Research Chemicals Supplier No. TRCT88630850MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3,5-Tri(1-naphthyl)benzene
CAS 7059-70-3
Molecular Formula C36H24
Quantity 50 mg
InChI Formula InChI=1S/C36H24/c1-4-16-31-25(10-1)13-7-19-34(31)28-22-29(35-20-8-14-26-11-2-5-17-32(26)35)24-30(23-28)36-21-9-15-27-12-3-6-18-33(27)36/h1-24H
SMILES c1ccc2c(cccc2c1)c3cc(cc(c3)c4cccc5ccccc45)c6cccc7ccccc67
IUPAC Name 1-(3,5-dinaphthalen-1-ylphenyl)naphthalene
Molecular Weight (g/mol) 456.58
Formula Weight 456.1878
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.