missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose

Product Code. 15490647
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490647 1 g 1g
15400657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490647 Supplier Thermo Scientific Chemicals Supplier No. J66805.03
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97614-43-2 | C26H21FO7 | 464.445 g/mol

It is a pharmaceutical intermediate and is used as an OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used as an OLED materials.

Solubility
Soluble in chloroform or dichloromethane

Notes
Refrigerate. Keep away from heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97614-43-2
Molecular Formula C26H21FO7
Molecular Weight (g/mol) 464.445
MDL Number MFCD00083339
InChI Key JOAHVPNLVYCSAN-UXGLMHHASA-N
Synonym 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-arabinofuranose, 2r,3s,4r,5r-5-benzoyloxy methyl-3-fluorotetrahydrofuran-2,4-diyl dibenzoate, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, a-d-arabinofuranose, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-a-d-arabinofuranose, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3s,4r,5r-5-benzoyloxymethyl-3-fluoro-tetrahydrofuran-2,4-diyl dibenzoate, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-md-arabinose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, pubchem20401, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-ribofuranose
PubChem CID 11754171
IUPAC Name [(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)F)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 76°C to 78°C
Color White
Quantity 1 g
Solubility Information Soluble in chloroform or dichloromethane
Formula Weight 464.44
Physical Form Powder
Chemical Name or Material 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.