missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triazine, 97%

Product Code. 11303836
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303836 1 g 1g
11313836 5 g 5g
11323836 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11303836 Supplier Thermo Scientific Alfa Aesar Supplier No. L16911.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 290-87-9 | C3H3N3 | 81.08 g/mol

1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

Solubility
Soluble in ethanol (50 mg/ml), and methanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 290-87-9
Molecular Formula C3H3N3
Molecular Weight (g/mol) 81.08
MDL Number MFCD00006044
InChI Key JIHQDMXYYFUGFV-UHFFFAOYSA-N
Synonym s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t
PubChem CID 9262
ChEBI CHEBI:30259
IUPAC Name 1,3,5-triazine
SMILES C1=NC=NC=N1

Specifications

Melting Point 77°C to 83°C (decomposition)
Density 1.38
Boiling Point 114°C
Quantity 1 g
Beilstein 104790
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol (50mg/ml),and methanol (100mg/ml).
Formula Weight 81.08
Percent Purity 97%
Chemical Name or Material 1,3,5-Triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.