Learn More
1,3,5-Triazine, 97%
Description
1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.
Solubility
Soluble in ethanol (50 mg/ml), and methanol (100 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under dry inert gas.
Specifications
Specifications
| Melting Point | 77°C to 83°C (decomposition) |
| Density | 1.38 |
| Boiling Point | 114°C |
| Quantity | 25 g |
| Beilstein | 104790 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in ethanol (50mg/ml),and methanol (100mg/ml). |
| Formula Weight | 81.08 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,3,5-Triazine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.