missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tribenzylhexahydro-1,3,5-triazine, 98+%

Product Code. 11445750
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445750 25 g 25g
11435750 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445750 Supplier Thermo Scientific Alfa Aesar Supplier No. H25964.14
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2547-66-2 | C24H27N3 | 357.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2547-66-2
Molecular Formula C24H27N3
Molecular Weight (g/mol) 357.50
MDL Number MFCD00014599
InChI Key VWVZIRPJPFJGFE-UHFFFAOYSA-N
Synonym 1,3,5-tribenzylhexahydro-1,3,5-triazine, 1,3,5-tribenzylhexahydro-s-triazine, 1,3,5-triazine, hexahydro-1,3,5-tris phenylmethyl, 1,3,5-trisbenzyl-1,3,5-triazaperhydroine, acmc-1cmek, 1,3,5-tribenzyl-1,3,5 triazinane, 1,3,5-triazine,hexahydro-1,3,5-tris phenylmethyl
PubChem CID 75685
IUPAC Name 1,3,5-tribenzyl-1,3,5-triazinane
SMILES C(N1CN(CC2=CC=CC=C2)CN(CC2=CC=CC=C2)C1)C1=CC=CC=C1

Specifications

Melting Point 50°C to 51°C
Boiling Point 100°C (0.005 mmHg)
Flash Point 113°C (235°F)
Quantity 25 g
Formula Weight 357.49
Percent Purity ≥98%
Chemical Name or Material 1,3,5-Tribenzylhexahydro-1,3,5-triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.