missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trichloro-2-nitrobenzene, 98+%

Product Code. 11424973
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424973 100 g 100g
11404973 5 g 5g
11414973 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11424973 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A12262.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18708-70-8 | C6H2Cl3NO2 | 226.437 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18708-70-8
Molecular Formula C6H2Cl3NO2
Molecular Weight (g/mol) 226.437
MDL Number MFCD00014690
InChI Key AEBJDOTVYMITIA-UHFFFAOYSA-N
PubChem CID 29228
IUPAC Name 1,3,5-trichloro-2-nitrobenzene
SMILES C1=C(C=C(C(=C1Cl)[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 68°C to 73°C
Quantity 100 g
UN Number UN2811
Beilstein 2213282
Synonym 2,4,6-trichloronitrobenzene, benzene, 1,3,5-trichloro-2-nitro, 2,4,6-trichloro-1-nitrobenzene, unii-t00kp6ct7k, t00kp6ct7k, benzene,1,3,5-trichloro-2-nitro, acmc-209epa, 2,6-trichloronitrobenzene, dsstox_cid_6204, dsstox_rid_78056
Formula Weight 226.45
Percent Purity ≥98%
Chemical Name or Material 1,3,5-Trichloro-2-nitrobenzene

RUO – Research Use Only

Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.