missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triethylbenzene, 95%

Product Code. 15477378
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477378 25 g 25g
15467378 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477378 Supplier Thermo Scientific Alfa Aesar Supplier No. H60875.14
 Restricted Item
Estimated Shipment: 05-01-2027
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 102-25-0 | C12H18 | 162.28 g/mol

,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.

Solubility
Insoluble in water. Soluble in alcohol, ether.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 102-25-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009261
InChI Key WJYMPXJVHNDZHD-UHFFFAOYSA-N
Synonym benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
PubChem CID 7602
IUPAC Name 1,3,5-triethylbenzene
SMILES CCC1=CC(CC)=CC(CC)=C1

Specifica

Density 0.862
Boiling Point 215°C
Flash Point 76°C (169°F)
Refractive Index 1.495
Quantity 25 g
UN Number UN3082
Beilstein 2039481
Solubility Information Insoluble in water. Soluble in alcohol,ether.
Formula Weight 162.27
Percent Purity 95%
Chemical Name or Material 1,3,5-Triethylbenzene
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 1,3,5-Triethylbenzene
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informazioni supplementari
  • MIXTURE LIST-Contain: 1,3,5-Triethylbenzene

RUO – Research Use Only

Titolo del prodotto
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.