missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trifluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Product Code. 11438896
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438896 25 g 25g
11418896 1 g 1g
11428896 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438896 Supplier Thermo Scientific Alfa Aesar Supplier No. A17035.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 315-14-0
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.082
MDL Number MFCD00014687
InChI Key PWRFDGYYJWQIAB-UHFFFAOYSA-N
Synonym 2,4,6-trifluoronitrobenzene, 2,2,6-trifluoronitrobenzene, benzene, 1,3,5-trifluoro-2-nitro, 1,3,5-trifluoro-2-nitro-benzene, 2-nitro-1,3,5-trifluorobenzene, pubchem10646, acmc-209hn4, 1,3,5-trifluoronitrobenzene, 2,4,6 trifluoronitrobenzene, ksc226o4t
PubChem CID 67567
IUPAC Name 1,3,5-trifluoro-2-nitrobenzene
SMILES C1=C(C=C(C(=C1F)[N+](=O)[O-])F)F

Specifications

Melting Point 2°C to 5°C
Density 1.514
Boiling Point 171°C to 172°C
Flash Point 77°C (170°F)
Refractive Index 1.479
Quantity 25 g
UN Number UN2810
Beilstein 2106869
Formula Weight 177.08
Percent Purity 98%
Chemical Name or Material 1,3,5-Trifluoro-2-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.