missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester, 95%

Product Code. 15448245
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448245 250 mg 250mg
15438245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448245 Supplier Thermo Scientific Alfa Aesar Supplier No. H32930.MD
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 844891-04-9 | C12H21BN2O2 | 236.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 844891-04-9
Molecular Formula C12H21BN2O2
Molecular Weight (g/mol) 236.12
MDL Number MFCD06659062
InChI Key IZNGYNMIIVJWSO-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3,5-trimethyl-1h-pyrazole-4-boronic acid pinacol ester, 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3,5-trimethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1,3,5-trimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3,5-trimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, pubchem19369, 1,3,5-trimethyl-1h-pyrazole-4-boronic pinacol ester, 1,3,5-trimethylpyrazole-4-boronic acid pinacol ester
PubChem CID 2795311
IUPAC Name 1,3,5-trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CN1N=C(C)C(B2OC(C)(C)C(C)(C)O2)=C1C

Specifications

Quantity 250 mg
Formula Weight 236.12
Percent Purity 95%
Chemical Name or Material 1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.