missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde, Thermo Scientific™

Product Code. p-4510138
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10101022 1 g Amber Glass Bottle 1g
10001364 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10101022 Supplier Thermo Scientific Maybridge Supplier No. CC14704DA
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2644-93-1
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
InChI Key HMTUBXVXHHITGO-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
PubChem CID 736515
IUPAC Name 1,3,5-trimethylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C)C)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.