missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde, 98%

Product Code. 11449340
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11449340 5 g 5g
11439340 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11449340

Brand: Thermo Scientific Alfa Aesar H50225.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2644-93-1 | C7H10N2O | 138.17 g/mol

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2644-93-1
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00159623
InChI Key HMTUBXVXHHITGO-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
PubChem CID 736515
IUPAC Name 1,3,5-trimethylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C)C)C=O

Specifications

Melting Point 80°C to 83°C
Boiling Point 132°C (12 mmHg)
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.