missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10716295
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10716295 10 g Amber Glass Bottle 10g
10436462 250 mg Amber Glass Bottle 250mg
10262682 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10716295 Supplier Thermo Scientific Maybridge Supplier No. CC14701EA
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1125-29-7
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00159648
InChI Key NOIOGQJFLIPRBI-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid
PubChem CID 736514
IUPAC Name 1,3,5-trimethylpyrazole-4-carboxylic acid
SMILES CN1N=C(C)C(C(O)=O)=C1C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.