missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(10-bromoanthracen-9-yl)benzene, TRC

Product Code. 30450982
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450982 50 mg 50mg
30453454 10 mg 10mg
30450969 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450982 Supplier Toronto Research Chemicals Supplier No. TRCT88650850MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813461-34-6
Molecular Formula C48H27Br3
Molecular Weight (g/mol) 843.44
Synonym 9,9',9' '-(1,3,5-benzenetriyl)tris[10-bromo-Anthracene, 9,9’,9’’-(1,3,5-Benzenetriyl)tris[10-bromoanthracene]
IUPAC Name 9-[3,5-bis(10-bromoanthracen-9-yl)phenyl]-10-bromoanthracene
SMILES Brc1c2ccccc2c(c3cc(cc(c3)c4c5ccccc5c(Br)c6ccccc46)c7c8ccccc8c(Br)c9ccccc79)c%10ccccc1%10

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C48H27Br3/c49-46-37-19-7-1-13-31(37)43(32-14-2-8-20-38(32)46)28-25-29(44-33-15-3-9-21-39(33)47(50)40-22-10-4-16-34(40)44)27-30(26-28)45-35-17-5-11-23-41(35)48(51)42-24-12-6-18-36(42)45/h1-27H
Formula Weight 839.9663
Chemical Name or Material 1,3,5-Tris(10-bromoanthracen-9-yl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.