missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminophenyl)ethanol, 98%

Product Code. 11382546
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382546 100 g 100g
11372546 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382546 Supplier Thermo Scientific Alfa Aesar Supplier No. L06513.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2454-37-7 | C8H11NO | 137.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2454-37-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007798
InChI Key QPKNDHZQPGMLCJ-UHFFFAOYSA-N
Synonym 1-3-aminophenyl ethanol, 3-1-hydroxyethyl aniline, m-1-hydroxyethyl aniline, m-amino-alpha-methylbenzyl alcohol, m-aminophenyl methyl carbinol, 1-3-aminophenyl ethan-1-ol, 3-amino-alpha-methylbenzyl alcohol, benzyl alcohol, m-amino-alpha-methyl, 3-aminophenyl-1-ethanol, benzyl alcohol, m-amino-.alpha.-methyl
PubChem CID 222461
IUPAC Name 1-(3-aminophenyl)ethanol
SMILES CC(C1=CC(=CC=C1)N)O

Specifications

Melting Point 66°C to 69°C
Boiling Point 217°C
Flash Point 157°C (314°F)
Quantity 100 g
Beilstein 2802415
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material 1-(3-Aminophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.