missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminopropyl)imidazole, 98%

Product Code. 11442217
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442217 250 g 250g
11432217 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442217 Supplier Thermo Scientific Alfa Aesar Supplier No. A14169.30
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5036-48-6 | C6H11N3 | 125.18 g/mol

1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents and air. Air sensitive. Store under dry inert gas. Incompatible with air and carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5036-48-6
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.18
MDL Number MFCD00009819
InChI Key KDHWOCLBMVSZPG-UHFFFAOYSA-N
Synonym 1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
PubChem CID 78736
SMILES NCCCN1C=CN=C1

Specifications

Melting Point -68°C
Density 1.062
pH 11.3
Boiling Point 296°C
Flash Point 154°C (309°F)
Odor Amine-like
Refractive Index 1.519
Quantity 250 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
IUPAC Name 3-imidazol-1-ylpropan-1-amine
Formula Weight 125.18
Percent Purity 98%
Chemical Name or Material 1-(3-Aminopropyl)imidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.