missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenediboronic acid, 97%, Thermo Scientific™

Product Code. 11411377
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411377 5 g 5g
11401377 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411377 Supplier Thermo Scientific Alfa Aesar Supplier No. B24903.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-benzenediboronic acid is used to prepare 1,3-didydrxybenzene by oxidation reaction using alkaline hydrogen peroxide.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4612-28-6
Molecular Formula C6H8B2O4
Molecular Weight (g/mol) 165.746
MDL Number MFCD03092915
InChI Key UXPAASVRXBULRG-UHFFFAOYSA-N
Synonym 1,3-benzenediboronic acid, 1,3-phenylenediboronic acid, 1,3-benzendiboronic acid, 1,3-benzenediboronicacid, m-phenylenediboronic acid, 3-dihydroxyboranyl phenyl boronic acid, 3-dihydroxyboranyl phenylboronic acid, m-benzenediboronic acid, pubchem17415, 1,3-benzendibororic acid
PubChem CID 230477
IUPAC Name (3-boronophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)B(O)O)(O)O

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 165.75
Percent Purity 97%
Chemical Name or Material 1,3-Benzenediboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.