missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedisulfonyl chloride, 97%

Product Code. 11486956
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486956 100 g 100g
11466956 5 g 5g
11476956 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486956 Supplier Thermo Scientific Alfa Aesar Supplier No. B24044.22
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 585-47-7 | C6H4Cl2O4S2 | 275.11 g/mol

1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

Solubility
Soluble in DMSO and methanol. Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-47-7
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.11
MDL Number MFCD00041879
InChI Key ALIQZUMMPOYCIS-UHFFFAOYSA-N
Synonym 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
PubChem CID 96240
SMILES ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 58°C to 60°C
Quantity 100 g
UN Number UN3261
Beilstein 1118508
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO and methanol. Reacts with water.
IUPAC Name benzene-1,3-disulfonyl chloride
Formula Weight 275.13
Percent Purity 98%
Chemical Name or Material 1,3-Benzenedisulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.