missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-4-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10506153
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10506153 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10506153 Supplier Thermo Scientific Maybridge Supplier No. CC01701CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5768-39-8
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD01076411
InChI Key DBUAYOWCIUQXQW-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxole-4-carboxylic acid, 2h-1,3-benzodioxole-4-carboxylic acid, 1,3-benzodioxole-4-carboxylicacid, 2,3-methylenedioxybenzoic acid, 2,3-methylenedioxy benzoic acid, o-piperonylic acid, 2-methylenedioxybenzoic acid, 4-carboxy-1,3-benzodioxole, 1-carboxy-methylenedioxybenzene, benzo 1,3 dioxole-4-carboxylic acid
PubChem CID 304832
IUPAC Name 1,3-benzodioxole-4-carboxylic acid
SMILES C1OC2=CC=CC(=C2O1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 166.13
Percent Purity 97%
Chemical Name or Material 1, 3-Benzodioxole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.