missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-5-carbonyl Chloride, TRC

Product Code. 30329812
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329812 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329812 Supplier Toronto Research Chemicals Supplier No. TRCB27672050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Benzodioxole-5-carbonyl Chloride
CAS 25054-53-9
Molecular Formula C8H5O3Cl
Quantity 50 mg
Synonym 2H-1,3-Benzodioxole-5-carbonyl Chloride, 2H-1,3-Benzodioxole-5-carboxylic Acid Chloride, 3,4-(Methylenedioxy)benzoyl Chloride, 3,4-Methylenedioxybenzoic Acid Chloride, Benzo[d][1,3]dioxole-5-carbonyl Chloride, Benzodioxole-5-carbonyl Chloride, Piperonylic Acid Chloride
InChI Formula InChI=1S/C8H5ClO3/c9-8(10)5-1-2-6-7(3-5)12-4-11-6/h1-3H,4H2
SMILES ClC(=O)c1ccc2OCOc2c1
IUPAC Name 1,3-benzodioxole-5-carbonyl chloride
Molecular Weight (g/mol) 184.57
Formula Weight 183.9927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.