missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole, 99%

Product Code. 10002120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10002120 50 g 50g
13438408 250 g 250g
13418408 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10002120 Supplier Thermo Scientific Acros Supplier No. 170220500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 274-09-9 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274-09-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00005818
InChI Key FTNJQNQLEGKTGD-UHFFFAOYSA-N
Synonym 1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene
PubChem CID 9229
ChEBI CHEBI:38732
IUPAC Name 1,3-benzodioxole
SMILES C1OC2=CC=CC=C2O1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -18.0°C
Color Brown to Yellow
Density 1.0600g/mL
Boiling Point 172.0°C to 173.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5380 to 1.5400
Quantity 50 g
Beilstein 19,2
Specific Gravity 1.06
Solubility Information Solubility in water: 0.2% (25 c). Other solubilities: miscible with most common organic solvents
Formula Weight 122.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 3-Benzodioxole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.