missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazol-2-ylmethylamine hydrochloride, ≥97%, Thermo Scientific™

Product Code. 10120632
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10120632 250 mg Amber Glass Bottle 250mg
10437992 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10120632 Supplier Thermo Scientific Maybridge Supplier No. CC06013CB
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 29198-41-2
Molecular Formula C8H9ClN2S
Molecular Weight (g/mol) 200.684
MDL Number MFCD03783535
InChI Key WCZDQDCFSDCIIC-UHFFFAOYSA-N
Synonym 1,3-benzothiazol-2-ylmethylamine hydrochloride, benzo d thiazol-2-ylmethanamine hydrochloride, 1,3-benzothiazol-2-ylmethanamine hydrochloride, 2-aminomethyl-1,3-benzothiazole hydrochloride, 1-1,3-benzothiazol-2-yl methanamine hydrochloride, 1,3-benzothiazol-2-yl-methanamine hydrochloride, c8h8n2s.clh, bestipharma 508-458, benzothiazol-2-ylmethylamine, chloride, benzothiazol-2-ylmethylamine hcl
PubChem CID 2776261
IUPAC Name 1,3-benzothiazol-2-ylmethanamine;hydrochloride
SMILES C1=CC=C2C(=C1)N=C(S2)CN.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.69
Percent Purity 95%
Chemical Name or Material 1, 3-Benzothiazol-2-ylmethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.