missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate, Thermo Scientific™

Product Code. 15467647
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467647 5 g 5g
15457647 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467647 Supplier Thermo Scientific Alfa Aesar Supplier No. H36330.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286014-42-4
Molecular Formula C23H33BF4N2
Molecular Weight (g/mol) 424.335
MDL Number MFCD04973311
InChI Key KVWCCJYLKCSVME-UHFFFAOYSA-N
Synonym 1,3-bis 1-adamantyl imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazolium tetrafluoroborate, 1,3-di adamantan-1-yl-1h-imidazol-3-ium tetrafluoroborate, 1,3-bis adamantan-1-yl-1??-imidazol-1-ylium tetrafluoroborate, iad.hbf4, acmc-209h3n, 1,3-diadamantyl-imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazoliumtetrafluoroborate, 1,3-bis tricyclo 3.3.1.13,7 dec-1-yl-1h-imidazolium tetrafluoroborate
PubChem CID 16218105
IUPAC Name 1,3-bis(1-adamantyl)imidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1C2CC3CC1CC(C2)(C3)N4C=C[N+](=C4)C56CC7CC(C5)CC(C7)C6

Specifications

Melting Point 280°C to 286°C
Color Brown to White
Quantity 5 g
Formula Weight 424.33
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.