missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, 97%

Product Code. 11456270
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456270 1 g 1g
11466270 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456270 Supplier Thermo Scientific Alfa Aesar Supplier No. H27150.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 250285-32-6 | C27H37ClN2 | 425.06 g/mol

1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride is also used in organic synthesis, as well as a pharmaceutical intermediate. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is used as a reagent in the synthesis of NHC Copper(I) complexes bearing dipyridylamine ligands which exhibit interesting luminescent properties and are potential candidates for organic light-emitting diode applications. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is also used as a reagent in the synthesis of 5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic Acid Methyl Ester (D476595); the methyl ester derivative of the drug Vadimezan (V084950).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride is also used in organic synthesis, as well as a pharmaceutical intermediate. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is used as a reagent in the synthesis of NHC Copper(I) complexes bearing dipyridylamine ligands which exhibit interesting luminescent properties and are potential candidates for organic light-emitting diode applications. 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride is also used as a reagent in the synthesis of 5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic Acid Methyl Ester (D476595); the methyl ester derivative of the drug Vadimezan (V084950).

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 250285-32-6
Molecular Formula C27H37ClN2
Molecular Weight (g/mol) 425.06
MDL Number MFCD02684545
InChI Key AVJBQMXODCVJCJ-UHFFFAOYSA-M
Synonym 1,3-bis 2,6-diisopropylphenyl imidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-1h-imidazol-3-ium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazolium chloride, ipr hcl, 2,5-bis 2,6-diisopropylphenyl imidazolium chloride, ipr.hcl, 1,3-bis 2,6-bis 1-methylethyl phenyl-1h-imidazolium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazoliumchloride, 1,3-bis-2,6-diisopropyl-phenyl-3h-imidazol-1-ium chloride, 1,3-bis 2,6-bis prop-2-yl phenyl-1h-imidazol-3-ium chloride
PubChem CID 2734913
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]imidazol-1-ium;chloride
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

Melting Point ∽280°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 425.05
Percent Purity 97%
Chemical Name or Material 1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride
compliance-icons
Product Identifier
  • 1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.