missing translation for 'onlineSavingsMsg'
Learn More

1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, 97%

Product Code. 10675903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
Packaging:
Glass bottle
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10675903 2 g Glass bottle 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10675903 Supplier Thermo Scientific Acros Supplier No. 378310020
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 250285-32-6 | C27H37ClN2 | 425.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 250285-32-6
CAS Min % 96.0
CAS Max % 100.0
Color Tan or White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C27H37ClN2
MDL Number MFCD02684545
Quantity 2 g
Synonym 1,3-bis 2,6-diisopropylphenyl imidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-1h-imidazol-3-ium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazolium chloride, ipr hcl, 2,5-bis 2,6-diisopropylphenyl imidazolium chloride, ipr.hcl, 1,3-bis 2,6-bis 1-methylethyl phenyl-1h-imidazolium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazoliumchloride, 1,3-bis-2,6-diisopropyl-phenyl-3h-imidazol-1-ium chloride, 1,3-bis 2,6-bis prop-2-yl phenyl-1h-imidazol-3-ium chloride
Solubility Information Solubility in water: slightly soluble
InChI Key AVJBQMXODCVJCJ-UHFFFAOYSA-M
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]imidazol-1-ium;chloride
Molecular Weight (g/mol) 425.06
PubChem CID 2734913
Formula Weight 425.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.