missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(2-chloroethyl-d4)-1-nitrosourea, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Carmustine-D8 (bis(2-chloroethyl-D4)) |
| Molecular Formula | C5 D8 H Cl2 N3 O2 |
| Quantity | 0.05 g |
| Synonym | 1,3-Bis(2-chloro-1,1,2,2-tetradeuterioethyl)-1-nitrosourea, N,N'-Bis(2-chloroethyl-1,1,2,2-d4)-N-nitrosourea, Carmustine-d8, Carmustine-D8 (bis(2-chloroethyl-D4)), 1,3-Bis(2-chloroethyl-d4)-1-nitrosourea |
| InChI Formula | InChI=1 S/C5H9Cl2N3O2/c6-1-3-8-5(11)10(9-12)4-2-7/h1-4H2,(H,8,11)/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])NC(=O)N(N=O)C([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | 1,3-bis(2-chloro-1,1,2,2-tetradeuterioethyl)-1-nitrosourea |
| Molecular Weight (g/mol) | 222.0992 |
| Formula Weight | 221.0574 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction