missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene, 96%

Product Code. 11333826
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333826 5 g 5g
11343826 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333826 Supplier Thermo Scientific Alfa Aesar Supplier No. L16894.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 802-93-7 | C12H6F12O2 | 410.159 g/mol

1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene is used to determine the specific details of electrical magnitudes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene is used to determine the specific details of electrical magnitudes.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 802-93-7
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.159
MDL Number MFCD00042090
InChI Key PGUIOHNOYADLMU-UHFFFAOYSA-N
Synonym 1,3-bis 2-hydroxyhexafluoroisopropyl benzene, 1,3-bis hexafluoro-2-hydroxyisopropyl benzene, 1,3-bis alpha-hydroxyhexafluoroisopropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,3-benzenedimethanol, a,a,a',a'-tetrakis trifluoromethyl-1,3-benzenedimethanol, 1,3-benzenedimethanol, .alpha.,.alpha.,.alpha.',.alpha.'-tetrakis trifluoromethyl, 1,3-bis hexafluoro-alpha-hydroxyisopropyl benzene, 1,1,1,3,3,3-hexafluoro-2-3-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,3-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, 1,3 hfab
PubChem CID 69933
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES C1=CC(=CC(=C1)C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O

Specifications

Melting Point 9°C to 10°C
Density 1.659
Boiling Point 99°C to 100°C (20 mmHg)
Flash Point 97°C (206°F)
Refractive Index 1.389
Quantity 5 g
Beilstein 2024879
Solubility Information Difficult to mix.
Formula Weight 410.16
Percent Purity 96%
Chemical Name or Material 1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.